(S)-(+)-3-Methyl-2-butanol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09431-MC
L09431-ME
CAAAAL09431-MCEA
81.53
CAD
CAAAAL09431-MC
CAAAAL09431-ME
(S)-(+)-3-Methyl-2-butanol 99%
(S)-(+)-3-Methyl-2-butanol
Purity: 99%
CAS No.: 1517-66-4
Molecular Formula: C5H12O
Formula Weight: 88.15
Flash Point: 26°(78°F)
Refractive Index: 1.4090
Boiling Point: 112°
Beilstein Reference: 1718801
Optical Rotation: +5° (neat)
UN No.: 1105
MDL No.: MFCD00064271
Formula:
C₅H₁₂O Boiling Pt: 112 °C Density: 0.810 Flash Pt: 26 °C (78 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064271 CAS Number: 1517-66-4 UN: 1105 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...