(S)-(-)-2-Chloropropionic acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L09427-06
L09427-14
CAAAAL09427-06EA
103.65
CAD
CAAAAL09427-06
CAAAAL09427-14
(S)-(-)-2-Chloropropionic acid 99%
(S)-(-)-2-Chloropropionic acid
Purity: 99%
CAS No.: 29617-66-1
Molecular Formula: C3H5ClO2
Formula Weight: 108.53
Flash Point: 107°(225°F)
Refractive Index: 1.4345
Melting Point: 4°
Boiling Point: 187°
Beilstein Reference: 1720257
UN No.: 2511
MDL No.: MFCD00064205
Formula:
H₃CCHClCOOH Boiling Pt: 187 °C Melting Pt: 4 °C Density: 1.270 Flash Pt: 107 °C (225 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064205 CAS Number: 29617-66-1 EINECS: 411-150-5 UN: 2511 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...