1-Tetradecanethiol 94%

Supplier: Thermo Scientific Chemicals

L09401-06 L09401-14
CAAAAL09401-06EA 61.15 CAD
CAAAAL09401-06 CAAAAL09401-14
1-Tetradecanethiol 94%
1-Tetradecanethiol

Purity: 94% CAS No.: 2079-95-0 Molecular Formula: C14H30S Formula Weight: 230.46 Flash Point: 110°(230°F) Refractive Index: 1.4610 Odor: Unpleasant Melting Point: 6-7° Boiling Point: 176-180°/22mm Beilstein Reference: 1742653 MDL No.: MFCD00022099

Formula: C₁₄H₃₀S
Boiling Pt: 176…180 °C/22 mm
Melting Pt: 6…7 °C
Density: 0.850
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00022099
CAS Number: 2079-95-0
EINECS: 218-209-2
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR