5-Chloro-2-nitrobenzamide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09380-04
L09380-09
CAAAAL09380-04EA
31.44
CAD
CAAAAL09380-04
CAAAAL09380-09
5-Chloro-2-nitrobenzamide 98+%
5-Chloro-2-nitrobenzamide
Purity: 98+%
CAS No.: 40763-96-0
Molecular Formula: C7H5ClN2O3
Formula Weight: 200.58
Melting Point: 155-159°
Beilstein Reference: 2267797
Formula:
C₇H₅ClN₂O₃ Melting Pt: 155…159 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017509 CAS Number: 40763-96-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...