4-Ethylcyclohexanone 99%
Supplier: Thermo Scientific Chemicals
Warning
|
L09363-14
L09363-22
CAAAAL09363-14EA
102.7
CAD
CAAAAL09363-14
CAAAAL09363-22
4-Ethylcyclohexanone 99%
4-Ethylcyclohexanone
Purity: 99%
CAS No.: 5441-51-0
Molecular Formula: C8H14O
Formula Weight: 126.20
Flash Point: 63°(145°F)
Refractive Index: 1.4515
Odor: Characteristic
Boiling Point: 192-194°
Beilstein Reference: 1924964
MDL No.: MFCD00001644
Formula:
C₈H₁₄O Boiling Pt: 192…194 °C Density: 0,895 Flash Pt: 63 °C(145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001644 CAS Number: 5441-51-0 EINECS: 226-629-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...