2-Pentadecanone 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Pentadecan-2-one

L09347-14 L09347-22
CAAAAL09347-14EA 79.63 CAD
CAAAAL09347-14 CAAAAL09347-22
2-Pentadecanone 97%
2-Pentadecanone

Purity: 97% CAS No.: 2345-28-0 Molecular Formula: C15H30O Formula Weight: 226.40 Flash Point: 144°(291°F) Melting Point: 37-41° Boiling Point: 293-294° Beilstein Reference: 1764872 MDL No.: MFCD00053712

Formula: C₁₅H₃₀O
MW: 226.4 g/mol
Boiling Pt: 293…294 °C
Melting Pt: 37…41 °C
Density: 0.82
Flash Pt: 144 °C(291 °F)
Storage Temperature: Ambient
MDL Number: MFCD00053712
CAS Number: 2345-28-0
EINECS: 219-064-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR