(±)-1-(1-Naphthyl)ethylamine 98%

Supplier: Thermo Scientific Chemicals
Warning

L09342-06 L09342-14
CAAAAL09342-06EA 78.84 CAD
CAAAAL09342-06 CAAAAL09342-14
(±)-1-(1-Naphthyl)ethylamine 98%
(±)-1-(1-Naphthyl)ethylamine

Purity: 98% CAS No.: 42882-31-5 Molecular Formula: C12H13N Formula Weight: 171.24 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.6230 Boiling Point: 155-156°/15mm Beilstein Reference: 3197375 UN No.: 2922 MDL No.: MFCD00004014

Formula: C₁₂H₁₃N
Boiling Pt: 155…156 °C (15 mmHg)
Density: 1.063
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004014
CAS Number: 42882-31-5
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR