Isopropyl acetate 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Acetic acid isopropyl ester
L09337-0F
L09337-AK
L09337-AU
CAAAAL09337-0FEA
188.02
CAD
CAAAAL09337-0F
CAAAAL09337-AK
CAAAAL09337-AU
Isopropyl acetate 99+%
Isopropyl acetate
Purity: 99+%
CAS No.: 108-21-4
Molecular Formula: C5H10O
Formula Weight: 102.13
Flash Point: 4°(39°F)
Refractive Index: 1.3770
Odor: Ester-like
Melting Point: -73°
Boiling Point: 87-89°
Beilstein Reference: 1740761
Merck Reference: 14,5205
UN No.: 1220
MDL No.: MFCD00008877
Formula:
C₅H₁₀O₂ Boiling Pt: 87…89 °C Melting Pt: –73 °C Density: 0.870 Flash Pt: 4 °C (39 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008877 CAS Number: 108-21-4 EINECS: 203-561-1 UN: 1220 ADR: 3,II Merck Index: 14,5205 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...