1-Adamantaneethanol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(2-Hydroxyethyl)adamantane, 1-(2-Hydroxyethyl)tricyclo[3.3.1.1(3,7)]decane, 2-(1-Adamantyl)ethanol

L09318-03 L09318-06
CAAAAL09318-03EA 75.21 CAD
CAAAAL09318-03 CAAAAL09318-06
1-Adamantaneethanol 98%
1-Adamantaneethanol

Purity: 98% CAS No.: 6240-11-5 Molecular Formula: C12H20O Formula Weight: 180.29 Melting Point: 73-75° Beilstein Reference: 1855382

Formula: C₁₂H₂₀O
MW: 180.29 g/mol
Boiling Pt: 270
Melting Pt: 73…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00074756
CAS Number: 6240-11-5
EINECS: 228-353-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR