4-Pentynoic acid 95%

Supplier: Thermo Scientific Chemicals
Danger

L09282-03 L09282-06
CAAAAL09282-03EA 101.28 CAD
CAAAAL09282-03 CAAAAL09282-06
4-Pentynoic acid 95%
4-Pentynoic acid

Purity: 98% CAS No.: 6089-09-4 Molecular Formula: C5H6O2 Formula Weight: 98.10 Storage Temperature: 2°C to 8°C Flash Point: 75°(167°F) Melting Point: 51-58° Boiling Point: 109-110°/30mm Beilstein Reference: 1742047 UN No.: 3261 MDL No.: MFCD00004407

Formula: C₅H₆O₂
Boiling Pt: 109…110 °C/30 mm
Melting Pt: 51…58 °C
Flash Pt: 75 °C (167 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004407
CAS Number: 6089-09-4
EINECS: 228-028-0
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR