4-(Methylthio)phenyl isothiocyanate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L09270-03
L09270-06
CAAAAL09270-03EA
44.24
CAD
CAAAAL09270-03
CAAAAL09270-06
4-(Methylthio)phenyl isothiocyanate 97%
4-(Methylthio)phenyl isothiocyanate
Purity: 97%
CAS No.: 15863-41-9
Molecular Formula: C8H7NS2
Formula Weight: 181.28
Flash Point:
Formula:
C₈H₇NS₂ Boiling Pt: 127…128 °C/1 mm Melting Pt: 48…51 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041368 CAS Number: 15863-41-9 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...