N,N,3,5-Tetramethylaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N,N-Dimethyl-3,5-xylidine, 3,5,N,N-Tetramethylaniline, 3,5-Dimethyl-N,N-dimethylaniline
L09266-06
L09266-14
CAAAAL09266-06EA
173.8
CAD
CAAAAL09266-06
CAAAAL09266-14
N,N,3,5-Tetramethylaniline 98%
N,N,3,5-Tetramethylaniline
Purity: 98%
CAS No.: 4913-13-7
Molecular Formula: C10H15N
Formula Weight: 149.24
Flash Point: 90°(194°F)
Refractive Index: 1.5440
Odor: Recognizable
Boiling Point: 226-228°
Beilstein Reference: 1933736
UN No.: 2810
MDL No.: MFCD00008307
Formula:
C₁₀H₁₅N MW: 149.24 g/mol Boiling Pt: 226…228 °C Density: 0.913 Flash Pt: 90 °C(194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008307 CAS Number: 4913-13-7 EINECS: 225-539-0 UN: 2810 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...