3-Fluorobenzoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Fluorobenzoylchloride, m-Fluorobenzoyl chloride
L09152-06
L09152-14
CAAAAL09152-06EA
47.08
CAD
CAAAAL09152-06
CAAAAL09152-14
3-Fluorobenzoyl chloride 98%
3-Fluorobenzoyl chloride
Purity: 98%
CAS No.: 1711-07-5
Molecular Formula: C7H4ClFO
Formula Weight: 158.56
Flash Point: 82°(179°F)
Refractive Index: 1.5285
Odor: Pungent
Melting Point: -30°
Boiling Point: 188-189°
Beilstein Reference: 636610
UN No.: 3265
MDL No.: MFCD00000670
Formula:
C₇H₄ClFO MW: 158.56 g/mol Boiling Pt: 188…189 °C Melting Pt: –30 °C Density: 1.320 Flash Pt: 82 °C (179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000670 CAS Number: 1711-07-5 EINECS: 216-977-3 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...