9-Bromophenanthrene 98%

Supplier: Thermo Scientific Chemicals

L09033-06 L09033-14
CAAAAL09033-06EA 49.14 CAD
CAAAAL09033-06 CAAAAL09033-14
9-Bromophenanthrene 98%
9-Bromophenanthrene

Purity: 98% CAS No.: 573-17-1 Molecular Formula: C14H9Br Formula Weight: 257.14 Melting Point: 62-66° Boiling Point: 190°/1.2mm MDL No.: MFCD00001174

Formula: C₁₄H₉Br
Boiling Pt: 190 °C/1.2 mm
Melting Pt: 62…66 °C
Density: 1.4093 g/cm³ (101 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001174
CAS Number: 573-17-1
EINECS: 209-351-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR