4-Nitro-m-anisic acid 98+%
Supplier: Thermo Scientific Chemicals
L08891-06
L08891-14
CAAAAL08891-06EA
53.09
CAD
CAAAAL08891-06
CAAAAL08891-14
4-Nitro-m-anisic acid 98+%
4-Nitro-m-anisic acid
Purity: 98+%
CAS No.: 5081-36-7
Molecular Formula: C8H7NO5
Formula Weight: 197.15
Melting Point: 231-235°
Beilstein Reference: 2696042
Formula:
C₈H₇NO₅ Boiling Pt: 405 °C (1013 hPa) Melting Pt: 231…235 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007353 CAS Number: 5081-36-7 EINECS: 225-793-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...