6-Methylnicotinic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L08877-03
L08877-06
L08877-14
CAAAAL08877-03EA
51.03
CAD
CAAAAL08877-03
CAAAAL08877-06
CAAAAL08877-14
6-Methylnicotinic acid 99%
6-Methylnicotinic acid
Purity: 99%
CAS No.: 3222-47-7
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Melting Point: 210-215°
Beilstein Reference: 112049
Formula:
C₇H₇NO₂ Melting Pt: 210…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006341 CAS Number: 3222-47-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...