4-Isopropylcyclohexanone 96%

Supplier: Thermo Scientific Chemicals
Warning

L08863-04 L08863-09
CAAAAL08863-04EA 135.56 CAD
CAAAAL08863-04 CAAAAL08863-09
4-Isopropylcyclohexanone 96%
4-Isopropylcyclohexanone

Purity: 96% CAS No.: 5432-85-9 Molecular Formula: C9H16O Formula Weight: 140.23 Flash Point: 80°(176°F) Refractive Index: 1.4580 Boiling Point: 94-96°/17mm Beilstein Reference: 1236952 MDL No.: MFCD00043479

Formula: C₉H₁₆O
MW: 140.23 g/mol
Boiling Pt: 94…96 °C (17 mmHg)
Flash Pt: 80 °C (176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00043479
CAS Number: 5432-85-9
EINECS: 226-592-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR