tert-Butyl acetate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L08855-0B
L08855-18
L08855-30
CAAAAL08855-0BEA
390.26
CAD
CAAAAL08855-0B
CAAAAL08855-18
CAAAAL08855-30
tert-Butyl acetate 99%
tert-Butyl acetate
Purity: 99%
CAS No.: 540-88-5
Molecular Formula: C6H12O2
Formula Weight: 116.16
Flash Point: 4°(39°F)
Refractive Index: 1.3865
Melting Point: -62°
Boiling Point: 94-96°
Beilstein Reference: 1699506
Merck Reference: 14,1537
UN No.: 1123
MDL No.: MFCD00008807
Formula:
C₆H₁₂O₂ Boiling Pt: 94…96 °C Melting Pt: –62 °C Density: 0.863 Flash Pt: 4 °C (39 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008807 CAS Number: 540-88-5 EINECS: 208-760-7 UN: 1123 ADR: 3,III Merck Index: 14,1537 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...