1,3-Diphenylpropane 98%
Supplier: Thermo Scientific Chemicals
L08849-04
L08849-09
CAAAAL08849-04EA
137.46
CAD
CAAAAL08849-04
CAAAAL08849-09
1,3-Diphenylpropane 98%
1,3-Diphenylpropane
Purity: 98%
CAS No.: 1081-75-0
Molecular Formula: C15H16
Formula Weight: 196.29
Refractive Index: 1.5600
Boiling Point: 158-159°/10mm
MDL No.: MFCD00043574
Formula:
(C₆H₅)CH₂CH₂CH₂(C₆H₅) Boiling Pt: 158…159 °C (10 mmHg) Density: 0.980 Storage Temperature: Ambient |
MDL Number:
MFCD00043574 CAS Number: 1081-75-0 EINECS: 214-101-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...