4-Bromo-2-methoxyphenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromoguaiacol, 5-Brom-2-hydroxyanisole
L08801-04
L08801-09
L08801-18
CAAAAL08801-04EA
63.04
CAD
CAAAAL08801-04
CAAAAL08801-09
CAAAAL08801-18
4-Bromo-2-methoxyphenol 98%
4-Bromo-2-methoxyphenol
Purity: 98%
CAS No.: 7368-78-7
Molecular Formula: C7H7BrO2
Formula Weight: 203.04
Odor: Phenol-like
Melting Point: 32-37°
Boiling Point: 129-132°/12mm
Beilstein Reference: 2045286
MDL No.: MFCD00051937
Formula:
C₇H₇BrO₂ MW: 203.04 g/mol Boiling Pt: 129…132 °C (12 mmHg) Melting Pt: 32…37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051937 CAS Number: 7368-78-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...