5α-Cholestan-3-one 97%
Supplier: Thermo Scientific Chemicals
L08726-03
L08726-06
CAAAAL08726-03EA
78.53
CAD
CAAAAL08726-03
CAAAAL08726-06
5α-Cholestan-3-one 97%
5α-Cholestan-3-one
Purity: 97%
CAS No.: 566-88-1
Molecular Formula: C27H46O
Formula Weight: 386.66
Melting Point: 128-132°
Beilstein Reference: 2625580
Formula:
C₂₇H₄₆O MW: 386.66 g/mol Melting Pt: 128…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065901 CAS Number: 566-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...