1,2-Dibenzoylethane 98+%
Supplier: Thermo Scientific Chemicals
L08652-03
L08652-06
CAAAAL08652-03EA
100.01
CAD
CAAAAL08652-03
CAAAAL08652-06
1,2-Dibenzoylethane 98+%
1,2-Dibenzoylethane
Purity: 98+%
CAS No.: 495-71-6
Molecular Formula: C16H14O2
Formula Weight: 238.29
Melting Point: 144-148°
Boiling Point: 260°/15mm
MDL No.: MFCD00037818
Formula:
C₁₆H₁₄O₂ Boiling Pt: 260 °C/15 mm Melting Pt: 144…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037818 CAS Number: 495-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...