4-Decylaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

L08620-03 L08620-06 L08620-14
CAAAAL08620-03EA 60.04 CAD
CAAAAL08620-03 CAAAAL08620-06 CAAAAL08620-14
4-Decylaniline 98%
4-Decylaniline

Purity: 98% CAS No.: 37529-30-9 Molecular Formula: C16H27N Formula Weight: 233.40 Flash Point: 110°(230°F) Refractive Index: 1.5100 Melting Point: 23-25° Boiling Point: 218-220°/14mm Beilstein Reference: 2721403 MDL No.: MFCD00007918

Formula: C₁₆H₂₇N
Boiling Pt: 218…220 °C/14 mm
Melting Pt: 23…25 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007918
CAS Number: 37529-30-9
EINECS: 253-546-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR