4-Chloro-m-toluic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L08533-04
L08533-09
L08533-18
CAAAAL08533-04EA
42.82
CAD
CAAAAL08533-04
CAAAAL08533-09
CAAAAL08533-18
4-Chloro-m-toluic acid 98%
4-Chloro-m-toluic acid
Purity: 98%
CAS No.: 7697-29-2
Molecular Formula: C8H7ClO2
Formula Weight: 170.60
Melting Point: 212-216°
Beilstein Reference: 1936535
Formula:
C₈H₇ClO₂ Melting Pt: 212…216 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045853 CAS Number: 7697-29-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...