4,5-Dimethoxy-2-nitrobenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L08510-06
L08510-14
L08510-22
CAAAAL08510-06EA
40.45
CAD
CAAAAL08510-06
CAAAAL08510-14
CAAAAL08510-22
4,5-Dimethoxy-2-nitrobenzoic acid 98%
4,5-Dimethoxy-2-nitrobenzoic acid
Purity: 98%
CAS No.: 4998-07-6
Molecular Formula: C9H9NO6
Formula Weight: 227.17
Melting Point: 192-195°
Beilstein Reference: 2123903
Formula:
C₉H₉NO₆ Melting Pt: 192…195 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014697 CAS Number: 4998-07-6 EINECS: 225-657-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...