3,5-Dimethylthiophenol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,3-Dimethyl-5-mercaptobenzene, 3,5-Dimethylbenzene-1-thiol, 3,5-Dimethylbenzenethiol, 5-Mercapto-m-xylene
L08403-06
L08403-14
CAAAAL08403-06EA
165.9
CAD
CAAAAL08403-06
CAAAAL08403-14
3,5-Dimethylthiophenol 97%
3,5-Dimethylthiophenol
Purity: 97%
CAS No.: 38360-81-5
Molecular Formula: C8H10S
Formula Weight: 138.23
Flash Point: 85°(185°F)
Refractive Index: 1.5675
Boiling Point: 127°/50mm
Beilstein Reference: 3233085
UN No.: 2810
MDL No.: MFCD00040226
Formula:
C₈H₁₀S MW: 138.23 g/mol Boiling Pt: 127 °C/50 mm Density: 1.015 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040226 CAS Number: 38360-81-5 EINECS: 253-901-8 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...