cis-6-Nonen-1-ol 95%
Supplier: Thermo Scientific Chemicals
L08387-03
L08387-06
CAAAAL08387-03EA
56.09
CAD
CAAAAL08387-03
CAAAAL08387-06
cis-6-Nonen-1-ol 95%
cis-6-Nonen-1-ol
Purity: 95%
CAS No.: 35854-86-5
Molecular Formula: C9H18O
Formula Weight: 142.24
Storage Temperature: 2°C to 8°C
Flash Point: 98°(208°F)
Refractive Index: 1.4510
Boiling Point: 85°/5mm
Beilstein Reference: 2322878
MDL No.: MFCD00015388
Formula:
C₉H₁₈O Boiling Pt: 85 °C/5 mm Density: 0.850 Flash Pt: 98 °C(208 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00015388 CAS Number: 35854-86-5 EINECS: 252-764-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...