2,3-Dimethylnaphthalene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L08386-03
L08386-06
CAAAAL08386-03EA
80.58
CAD
CAAAAL08386-03
CAAAAL08386-06
2,3-Dimethylnaphthalene 97%
2,3-Dimethylnaphthalene
Purity: 97%
CAS No.: 581-40-8
Molecular Formula: C12H12
Formula Weight: 156.23
Melting Point: 103-106°
Boiling Point: 265-268°
MDL No.: MFCD00004119
Formula:
C₁₂H₁₂ Boiling Pt: 265…268 °C Melting Pt: 103…106 °C Density: 1.003 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004119 CAS Number: 581-40-8 EINECS: 209-463-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...