Methyl-4-cyanobenzoate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L08365-06
L08365-14
L08365-22
CAAAAL08365-06EA
64.46
CAD
CAAAAL08365-06
CAAAAL08365-14
CAAAAL08365-22
Methyl-4-cyanobenzoate 99%
Methyl 4-cyanobenzoate
Purity: 99%
CAS No.: 1129-35-7
Molecular Formula: C9H7NO2
Formula Weight: 161.16
Melting Point: 65-69°
Boiling Point: 142-144°/12mm
MDL No.: MFCD00001823
Formula:
C₉H₇NO₂ Boiling Pt: 142…144 °C/12 mm Melting Pt: 65…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001823 CAS Number: 1129-35-7 EINECS: 214-443-4 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...