2,4-Dimethylquinoline 95%

Supplier: Thermo Scientific Chemicals
Warning

L08348-MC L08348-ME
CAAAAL08348-MCEA 43.61 CAD
CAAAAL08348-MC CAAAAL08348-ME
2,4-Dimethylquinoline 95%
2,4-Dimethylquinoline

Purity: 95% CAS No.: 1198-37-4 Molecular Formula: C11H11N Formula Weight: 157.21 Flash Point: 110°(230°F) Refractive Index: 1.6050 Boiling Point: 264-265° Beilstein Reference: 114230 MDL No.: MFCD00006760

Formula: C₁₁H₁₁N
Boiling Pt: 264…265 °C
Density: 1.060
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006760
CAS Number: 1198-37-4
EINECS: 214-832-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR