Azulene 99%

Supplier: Thermo Scientific Chemicals

L08271-03 L08271-MD
CAAAAL08271-03EA 338.12 CAD
CAAAAL08271-03 CAAAAL08271-MD
Azulene 99%
Azulene

Purity: 99% CAS No.: 275-51-4 Molecular Formula: C10H8 Formula Weight: 128.17 Melting Point: 98-101° Boiling Point: 241-243° Merck Reference: 14,926 MDL No.: MFCD00003810

Formula: C₁₀H₈
Boiling Pt: 241…243 °C
Melting Pt: 98…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00003810
CAS Number: 275-51-4
EINECS: 205-993-6
ADR: 9,III
Merck Index: 14,926

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR