D-(-)-Norleucine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-D-Nle-OH, (R)-(-)-Norleucine

L08257-03 L08257-06
CAAAAL08257-03EA 85.32 CAD
CAAAAL08257-03 CAAAAL08257-06
D-(-)-Norleucine 99%
D(-)-Norleucine

Purity: 99% CAS No.: 327-56-0 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Melting Point: 300° Beilstein Reference: 1721749 Merck Reference: 14,6706 Optical Rotation: -23° (c=5 in 5N HCl) MDL No.: MFCD00008099

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00008099
CAS Number: 327-56-0
EINECS: 206-320-9
Merck Index: 14,6706

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR