2-Amino-5-hydroxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Hydroxyanthranilic acid

L08256-06 L08256-14
CAAAAL08256-06EA 58.46 CAD
CAAAAL08256-06 CAAAAL08256-14
2-Amino-5-hydroxybenzoic acid 98%
2-Amino-5-hydroxybenzoic acid

Purity: 98% CAS No.: 394-31-0 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Melting Point: ca 248° dec. Beilstein Reference: 2803663

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: ∼ 248 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007870
CAS Number: 394-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR