2-Chloro-4-nitrobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Chloro-4-nitrobenzoylchloride, 2-Chloro-4-nitrobenzene-1-carbonyl chloride

L08246-06 L08246-18
CAAAAL08246-06EA 78.68 CAD
CAAAAL08246-06 CAAAAL08246-18
2-Chloro-4-nitrobenzoyl chloride 98%
2-Chloro-4-nitrobenzoyl chloride

Purity: 98% CAS No.: 7073-36-1 Molecular Formula: C7H3Cl2NO3 Formula Weight: 220.01 Refractive Index: 1.5950 Boiling Point: 165°/20mm UN No.: 3265 MDL No.: MFCD00051515

Formula: C₇H₃Cl₂NO₃
MW: 220.01 g/mol
Boiling Pt: 165 °C (20 mmHg)
Melting Pt: 115
Storage Temperature: Ambient
MDL Number: MFCD00051515
CAS Number: 7073-36-1
EINECS: 230-367-4
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR