4,4'-Dibromobenzophenone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Bis(4-bromophenyl) ketone
L08200-03
L08200-06
CAAAAL08200-03EA
123.24
CAD
CAAAAL08200-03
CAAAAL08200-06
4,4'-Dibromobenzophenone 98+%
4,4'-Dibromobenzophenone
Purity: 98+%
CAS No.: 3988-03-2
Molecular Formula: C13H8Br2O
Formula Weight: 340.03
Melting Point: 172-176°
Beilstein Reference: 2049958
Formula:
C₁₃H₈Br₂O MW: 340.01 g/mol Melting Pt: 172…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016330 CAS Number: 3988-03-2 EINECS: 223-632-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...