3,4-Difluorobenzoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L08188-06
L08188-14
CAAAAL08188-06EA
98.12
CAD
CAAAAL08188-06
CAAAAL08188-14
3,4-Difluorobenzoyl chloride 98%
3,4-Difluorobenzoyl chloride
Purity: 98%
CAS No.: 76903-88-3
Molecular Formula: C7H3ClF2O
Formula Weight: 176.55
Flash Point: 78°(172°F)
Refractive Index: 1.5110
Boiling Point: 185-186°
Beilstein Reference: 4177619
UN No.: 3265
MDL No.: MFCD00009930
Formula:
C₇H₃ClF₂O Boiling Pt: 185…186 °C Density: 1.402 Flash Pt: 78 °C (172 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009930 CAS Number: 76903-88-3 EINECS: 278-570-7 UN: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...