2,3,4-Trimethylpentane 98+%
Supplier: Thermo Scientific Chemicals
L08176-06
L08176-14
CAAAAL08176-06EA
104.12
CAD
CAAAAL08176-06
CAAAAL08176-14
2,3,4-Trimethylpentane 98+%
2,3,4-Trimethylpentane
Purity: 98+%
CAS No.: 565-75-3
Molecular Formula: C8H18
Formula Weight: 114.23
Flash Point: 5°(41°F)
Refractive Index: 1.4040
Melting Point: -110°
Boiling Point: 113-115°
Beilstein Reference: 1696869
UN No.: 1262
MDL No.: MFCD00008924
Formula:
C₈H₁₈ |
MDL Number:
MFCD00008924 CAS Number: 565-75-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...