Tetrabutylammonium hydroxide 1.0 M in methanol

Supplier: Thermo Scientific Chemicals
Danger

L08164-AC L08164-AE L08164-AP
CAAAAL08164-ACEA 62.41 CAD
CAAAAL08164-AC CAAAAL08164-AE CAAAAL08164-AP
Tetrabutylammonium hydroxide 1.0 M in methanol
Tetrabutylammonium hydroxide

CAS No.: 2052-49-5 Molecular Formula: [CH3(CH2)3]4NOH Formula Weight: 259.48 Flash Point: 11°(51°F) Refractive Index: 1.3825 Odor: Alcohol-like Beilstein Reference: 3571228 UN No.: 2924 MDL No.: MFCD00009425

Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
Density: 0.830
Flash Pt: 11 °C (51 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 3286
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR