3-(Trifluoromethyl)benzyl mercaptan tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(Trifluoromethyl)benzylthiol

L08155-03 L08155-06
CAAAAL08155-03EA 81.53 CAD
CAAAAL08155-03 CAAAAL08155-06
3-(Trifluoromethyl)benzyl mercaptan tech. 90%, Technical Grade
3-(Trifluoromethyl)benzyl mercaptan

Purity: 90% CAS No.: 25697-55-6 Molecular Formula: C8H7F3S Formula Weight: 192.20 Refractive Index: 1.4900 Boiling Point: 75-77°/5mm UN No.: 2810 MDL No.: MFCD00042435

Formula: C₈H₇F₃S
MW: 192.2 g/mol
Boiling Pt: 75…77 °C (5 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00042435
CAS Number: 25697-55-6
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR