Triphenyl phosphate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L08130-0B
L08130-30
CAAAAL08130-0BEA
151.68
CAD
CAAAAL08130-0B
CAAAAL08130-30
Triphenyl phosphate 98%
Triphenyl phosphate
Purity: 98%
CAS No.: 115-86-6
Molecular Formula: C18H15O4P
Formula Weight: 326.29
Flash Point: 223°(433°F)
Odor: Odorless
Boiling Point: 247-250°/11mm
Beilstein Reference: 1888236
Merck Reference: 14,9742
UN No.: 3077
MDL No.: MFCD00003031
Formula:
C₁₈H₁₅O₄P Boiling Pt: 247…250 °C/11 mm Melting Pt: 48…52 °C Density: 1.33 g/cm³ (20 °C) Flash Pt: 223 °C (433 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003031 CAS Number: 115-86-6 EINECS: 204-112-2 UN: 3077 ADR: 9,III Merck Index: 14,9742 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...