trans-2-Methyl-2-butenal 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tiglic aldehyde, (E)-2-Methyl-2-butenal
L08113-06
L08113-14
CAAAAL08113-06EA
74.73
CAD
CAAAAL08113-06
CAAAAL08113-14
trans-2-Methyl-2-butenal 97%
trans-2-Methyl-2-butenal
Purity: 97%
CAS No.: 497-03-0
Molecular Formula: C5H8O
Formula Weight: 84.12
Storage Temperature: 2°C to 8°C
Flash Point: 18°(64°F)
Refractive Index: 1.4480
Odor: Fruit-like
Boiling Point: 117-119°
Beilstein Reference: 1698207
UN No.: 1989
MDL No.: MFCD00006977
Formula:
C₅H₈O MW: 84.12 g/mol Boiling Pt: 117…119 °C Density: 0.871 Flash Pt: 18 °C (64 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00006977 CAS Number: 497-03-0 EINECS: 207-833-0 UN: 1989 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...