2-Methoxy-3-methylpyrazine 99%

Supplier: Thermo Scientific Chemicals
Warning

L08067-06 L08067-14
CAAAAL08067-06EA 93.69 CAD
CAAAAL08067-06 CAAAAL08067-14
2-Methoxy-3-methylpyrazine 99%
2-Methoxy-3-methylpyrazine

Purity: 99% CAS No.: 2847-30-5 Molecular Formula: C6H8N2O Formula Weight: 124.14 Flash Point: 55°(131°F) Refractive Index: 1.5065 Boiling Point: 159° Beilstein Reference: 878809 UN No.: 1993 MDL No.: MFCD00006127

Formula: C₆H₈N₂O
MW: 124.14 g/mol
Boiling Pt: 159 °C
Density: 1.070
Flash Pt: 55 °C (131 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006127
CAS Number: 2847-30-5
EINECS: 220-651-6
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR