(±)-2-Methyl-3-pentanol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L08015-09
L08015-18
CAAAAL08015-09EA
145.04
CAD
CAAAAL08015-09
CAAAAL08015-18
(±)-2-Methyl-3-pentanol 99%
(±)-2-Methyl-3-pentanol
Purity: 99%
CAS No.: 565-67-3
Molecular Formula: C6H14O
Formula Weight: 102.18
Flash Point: 46°(114°F)
Refractive Index: 1.4170
Boiling Point: 127-129°
Beilstein Reference: 1718952
UN No.: 2282
MDL No.: MFCD00004567
Formula:
C₆H₁₄O Boiling Pt: 127…129 °C Melting Pt: –53 °C Density: 0.830 Flash Pt: 46 °C (114 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004567 CAS Number: 565-67-3 EINECS: 209-286-3 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...