5-Methylisatin 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L08009-03
L08009-06
L08009-14
CAAAAL08009-03EA
53.88
CAD
CAAAAL08009-03
CAAAAL08009-06
CAAAAL08009-14
5-Methylisatin 97%
5-Methylisatin
Purity: 97%
CAS No.: 608-05-9
Molecular Formula: C9H7NO2
Formula Weight: 161.16
Melting Point: ca 180° dec.
Beilstein Reference: 123738
Formula:
C₉H₇NO₂ Melting Pt: ∼ 180 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005721 CAS Number: 608-05-9 EINECS: 210-152-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...