7-Azaindole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Pyrrolo[2,3-b]pyridine

L07983-03 L07983-06 L07983-14
CAAAAL07983-03EA 43.77 CAD
CAAAAL07983-03 CAAAAL07983-06 CAAAAL07983-14
7-Azaindole 98%
7-Azaindole

Purity: 98% CAS No.: 271-63-6 Molecular Formula: C7H6N2 Formula Weight: 118.14 Melting Point: 103-107° Boiling Point: 270° MDL No.: MFCD00005606

Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 270 °C
Melting Pt: 103…107 °C
Density: 1.301 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005606
CAS Number: 271-63-6
EINECS: 205-981-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR