7-Nitro-1H-indazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
7-Nitroindazole, 7-NINA, 7-Nitro-1H-indazole
L07970-03
L07970-06
L07970-14
CAAAAL07970-03EA
135.56
CAD
CAAAAL07970-03
CAAAAL07970-06
CAAAAL07970-14
7-Nitro-1H-indazole 98%
7-Nitro-1H-indazole
Nitrix oxide synthase inhibitor
Purity: 98%
CAS No.: 2942-42-9
Molecular Formula: C7H5N3O2
Formula Weight: 163.14
Melting Point: 184-189°
Beilstein Reference: 6809
Formula:
C₇H₅N₃O₂ MW: 163.14 g/mol Melting Pt: 190 Storage Temperature: Ambient |
MDL Number:
MFCD00022789 CAS Number: 2942-42-9 EINECS: 220-934-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...