Linoleic acid 95%
Supplier: Thermo Scientific Chemicals
L07949-06
L07949-14
L07949-22
CAAAAL07949-06EA
104.12
CAD
CAAAAL07949-06
CAAAAL07949-14
CAAAAL07949-22
Linoleic acid 95%
Linoleic acid
Unsaturated omega-6 fatty acid
Purity: 95%
CAS No.: 60-33-3
Molecular Formula: C18H32O2
Formula Weight: 280.45
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₁₈H₃₂O₂ Boiling Pt: 230 °C/16 mm Melting Pt: –12 °C Density: 0.902 Flash Pt: >110 °C(230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00064241 CAS Number: 60-33-3 EINECS: 200-470-9 Merck Index: 14,5505 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...