(±)-3-(Diethylamino)-1,2-propanediol 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L07943-14
L07943-22
L07943-36
CAAAAL07943-14EA
46.77
CAD
CAAAAL07943-14
CAAAAL07943-22
CAAAAL07943-36
(±)-3-(Diethylamino)-1,2-propanediol 97%
(±)-3-(Diethylamino)-1,2-propanediol
Purity: 97%
CAS No.: 621-56-7
Molecular Formula: C7H17NO2
Formula Weight: 147.22
Flash Point: 107°(224°F)
Refractive Index: 1.4602
Boiling Point: 233-235°
Beilstein Reference: 1737402
UN No.: 2735
MDL No.: MFCD00004716
Formula:
C₇H₁₇NO₂ Boiling Pt: 233…235 °C Melting Pt: –30 °C Density: 0.970 Flash Pt: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004716 CAS Number: 621-56-7 EINECS: 210-693-3 UN: 3267 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...