2-Phenylpyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

L07861-09 L07861-18
CAAAAL07861-09EA 106.18 CAD
CAAAAL07861-09 CAAAAL07861-18
2-Phenylpyridine 98%
2-Phenylpyridine

Purity: 98% CAS No.: 1008-89-5 Molecular Formula: C11H9N Formula Weight: 155.20 Flash Point: 110°(230°F) Refractive Index: 1.6240 Melting Point: -5° Boiling Point: 268-270° Beilstein Reference: 110445 MDL No.: MFCD00006280

Formula: C₁₁H₉N
Boiling Pt: 268…270 °C
Melting Pt: –5 °C
Density: 1.086
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006280
CAS Number: 1008-89-5
EINECS: 213-763-1
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR