3-Aminobenzyl alcohol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L07847-06
L07847-14
CAAAAL07847-06EA
87.85
CAD
CAAAAL07847-06
CAAAAL07847-14
3-Aminobenzyl alcohol 98+%
3-Aminobenzyl alcohol
Purity: 98+%
CAS No.: 1877-77-6
Molecular Formula: C7H9NO
Formula Weight: 123.16
Storage Temperature: 2°C to 8°C
Melting Point: 91-95°
Beilstein Reference: 2205844
MDL No.: MFCD00007817
Formula:
C₇H₉NO Boiling Pt: 272 °C (1013 hPa) Melting Pt: 91…95 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007817 CAS Number: 1877-77-6 EINECS: 217-514-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...